Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Penicillin-Streptomycin-Glutamine (100X)
GREENER_CHOICE

Catalog No. 10378016
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-378-016 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-378-016 Supplier Gibco™ Supplier No. 10378016
Only null left
Add to Cart
Add to Cart

For prevention from bacterial contamination in cell cultures due to their effective combined action against gram-positive and gram-negative bacteria.

Penicillin was originally purified from the fungi Penicillium and acts by interfering directly with the turnover of the bacteria cell wall and indirectly by triggering the release of enzymes that alter cell wall. Streptomycin was originally purified from Streptomyces griseus. It acts by binding to 30S subunit of the bacterial ribosome, leading to the inhibition of protein synthesis and death in susceptible bacteria.

  • Available as 100 mL liquid
  • Solution contains 10,000 units of penicillin, 10,000μg of streptomycin, and 29.2 mg/mL of L-glutamine in a 10 mM citrate buffer (for pH stability)

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: -5°C to -20°C
Shipping conditions: Dry ice
Shelf life: 12 months from date of manufacture
Concentration 100 X, 50 mg/mL
Form Liquid
Product Type Antibiotic-L-Glutamine
Sterility Sterile-filtered
With Additives Glutamine
For Use With (Application) Prevention of Cell Culture Contamination
Quantity 100 mL
Shipping Condition Dry Ice
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.