Learn More
Medchemexpress LLC HY-116790A 10mg Medchemexpress, (+)-Penbutolol CAS:38363-41-6 Purity:>98%
SDP
Catalog No. 501969440
Encompass_Preferred
Medchemexpress, HY-116790A 10mg (+)-Penbutolol CAS:38363-41-6 (+)-Penbutolol is a β-adrenoceptor antagonist, with an IC50 of 0.74 μM. (+)-Penbutolol is an optical isomer of l-penbutolol with Na+ channel-blocking action. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9440 Supplier Medchemexpress LLC Supplier No. HY116790A10MG
Medchemexpress, HY-116790A 10mg (+)-Penbutolol CAS:38363-41-6 (+)-Penbutolol is a β-adrenoceptor antagonist, with an IC50 of 0.74 μM. (+)-Penbutolol is an optical isomer of l-penbutolol with Na+ channel-blocking action. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.