Learn More
Medchemexpress LLC Penbutolol sulfate | 38363-32-5 | 98.9% | 50 MG
SDP
Catalog No. 5000413100
Encompass_Preferred
Penbutolol sulfate, also known as (-)-Terbuclomine, is a potent antagonist for both β-adrenoceptor and 5-HT receptors. It has demonstrated Ki values of 11.6 nM and 11.9 nM for 5-HT in rat cornu ammonis 1 (CA1) and human CA3, respectively. This product is intended strictly for research purposes.
  • Acts as a potent antagonist for β-adrenoceptor and 5-HT receptors.
  • Exhibits specific Ki values for 5-HT in CA1 and CA3.
  • Suitable for various research applications.
Catalog No. 50-004-13100 Supplier Medchemexpress LLC Supplier No. HYB115450MG
Penbutolol sulfate, also known as (-)-Terbuclomine, is a potent antagonist for both β-adrenoceptor and 5-HT receptors. It has demonstrated Ki values of 11.6 nM and 11.9 nM for 5-HT in rat cornu ammonis 1 (CA1) and human CA3, respectively. This product is intended strictly for research purposes.
  • Acts as a potent antagonist for β-adrenoceptor and 5-HT receptors.
  • Exhibits specific Ki values for 5-HT in CA1 and CA3.
  • Suitable for various research applications.