Learn More
TARGETMOL CHEMICALS INC PENBUTOLOL SULFATE 50MG
SDP
Catalog No. 502577425
Encompass_Preferred
Also available in 10 mg 100 mg and bulk. Please contact Fisher for quotes. Penbutolol sulfate ((-)-Terbuclomine) is able to bind to both (beta)2-adrenergic receptor and (beta)1-adrenergic receptor thus making it a non-selective (beta) blocker. purity: 98%
Catalog No. 50-257-7425 Supplier TARGETMOL CHEMICALS INC Supplier No. T1240250MG
Also available in 10 mg 100 mg and bulk. Please contact Fisher for quotes. Penbutolol sulfate ((-)-Terbuclomine) is able to bind to both (beta)2-adrenergic receptor and (beta)1-adrenergic receptor thus making it a non-selective (beta) blocker. purity: 98%