Learn More
Medchemexpress LLC Penbutolol sulfate ((-)-Terbuclomine) | 38363-32-5 | 98.89% | 5 MG
SDP
Catalog No. 5000411345
Encompass_Preferred
Penbutolol sulfate, also known as (-)-Terbuclomine, is a potent antagonist targeting both β-adrenoceptors and 5-HT receptors. It exhibits high affinity for these receptors, with reported Ki values of 11.6 nM for β-adrenoceptors and 11.9 nM for 5-HT receptors in specific rat and human brain regions. This compound is intended for research applications only.
  • Potent β-adrenoceptor and 5-HT receptor antagonist.
  • Ki values of 11.6 nM for β-adrenoceptors and 11.9 nM for 5-HT receptors.
  • Inhibits 5-HT cell discharge in rat models.
  • Enhances the effect of Paroxetine on elevating 5-HT levels in rat models.
Catalog No. 50-004-11345 Supplier Medchemexpress LLC Supplier No. HYB11545MG
Penbutolol sulfate, also known as (-)-Terbuclomine, is a potent antagonist targeting both β-adrenoceptors and 5-HT receptors. It exhibits high affinity for these receptors, with reported Ki values of 11.6 nM for β-adrenoceptors and 11.9 nM for 5-HT receptors in specific rat and human brain regions. This compound is intended for research applications only.
  • Potent β-adrenoceptor and 5-HT receptor antagonist.
  • Ki values of 11.6 nM for β-adrenoceptors and 11.9 nM for 5-HT receptors.
  • Inhibits 5-HT cell discharge in rat models.
  • Enhances the effect of Paroxetine on elevating 5-HT levels in rat models.