Learn More
Medchemexpress LLC HY-17424 50mg Medchemexpress, Penciclovir CAS:39809-25-1 Purity:>98%
SDP
Catalog No. 502081949
Encompass_Preferred
Medchemexpress, HY-17424 50mg Penciclovir CAS:39809-25-1 Penciclovir is reported to be potent against HSV types 1 and 2 with IC50 of 0.04-1.8 μg/mL and 0.06-4.4 μg/mL, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1949 Supplier Medchemexpress LLC Supplier No. HY1742450MG
Medchemexpress, HY-17424 50mg Penciclovir CAS:39809-25-1 Penciclovir is reported to be potent against HSV types 1 and 2 with IC50 of 0.04-1.8 μg/mL and 0.06-4.4 μg/mL, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.