Learn More
Research Products International Corp PENICILLIN/STREPTOMYCIN/NEOMYCIN MIXTURE, 50ML
SDP
Catalog No. 501368544
Encompass_Preferred
Penicillin, Streptomycin, and Neomycin solution is a common combination antibiotic mixture that inhibits the growth of bacteria. This specific combination creates a broad-spectrum antibiotic effective against a wide range of Gram-positive and Gram-negative bacteria. 
This soluion is commonly used in cell culture and microbiology research to prevent bacterial contamination. When added to a cell culture medium, it creates a sterile environment unfavorable for bacterial growth.
WARNING! This product can expose you to Streptomycin sulfate and Neomycin sulfate, chemicals which are known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-136-8544 Supplier Research Products International Corp Supplier No. P9358050.0
Penicillin, Streptomycin, and Neomycin solution is a common combination antibiotic mixture that inhibits the growth of bacteria. This specific combination creates a broad-spectrum antibiotic effective against a wide range of Gram-positive and Gram-negative bacteria. 
This soluion is commonly used in cell culture and microbiology research to prevent bacterial contamination. When added to a cell culture medium, it creates a sterile environment unfavorable for bacterial growth.
WARNING! This product can expose you to Streptomycin sulfate and Neomycin sulfate, chemicals which are known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.