Learn More
Medchemexpress LLC HY-135761 100mg Medchemexpress, Penconazole CAS:66246-88-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY135761100MG
Encompass_Preferred
Medchemexpress, HY-135761 100mg Penconazole CAS:66246-88-6 Penconazole is a typical triazole fungicide, and mainly applied on apples, grapes, and vegetables to control powdery mildew. Penconazole inhibits sterol biosynthesis in fungi. Penconazole decrease AChE activity in the cerebrum and cerebellum of rats. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1400