Learn More
Medchemexpress LLC Pencycuron-d5 | 83286-58-2 | 99.90% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB2048S1MG
Encompass_Preferred
Pencycuron-d5 is a deuterium-labeled version of Pencycuron, identified as a benzoylurea fungicide. Its fungicidal action involves inhibiting the synthesis of fungal cell walls. This compound can be utilized in research focused on controlling fungal diseases in crops, including conditions such as sheath blight, powdery mildew, and late blight.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-12299