Learn More
Apexbio Technology LLC PENDIMETHALIN-200MG
SDP
Catalog No. 5000568817
Encompass_Preferred
Pendimethalin (CAS No 40487-42-1) is a pre-emergent herbicide widely used in agricultural research to study its effects on weed control Originating from the dinitroaniline chemical family its primary mechanism of action involves the inhibition of microtubule assembly preventing cell division in target plant cells In vitro studies have demonstrated that its activity generally falls within nanomolar to low micromolar ranges making it effective in disrupting cellular processes at relatively low concentrations Additionally Pendimethalin is commonly utilized in various cell models and animal studies to evaluate its efficacy and toxicity These studies provide insight into its potential environmental impact and guide the development of new agrochemical formulations Experimental concentrations typically depend on the specific research design Overall Pendimethalin remains a critical compound in herbicide research contributing to advancements in sustainable agricultural practices
Catalog No. 50-005-68817 Supplier Apexbio Technology LLC Supplier No. C6572200
Pendimethalin (CAS No 40487-42-1) is a pre-emergent herbicide widely used in agricultural research to study its effects on weed control Originating from the dinitroaniline chemical family its primary mechanism of action involves the inhibition of microtubule assembly preventing cell division in target plant cells In vitro studies have demonstrated that its activity generally falls within nanomolar to low micromolar ranges making it effective in disrupting cellular processes at relatively low concentrations Additionally Pendimethalin is commonly utilized in various cell models and animal studies to evaluate its efficacy and toxicity These studies provide insight into its potential environmental impact and guide the development of new agrochemical formulations Experimental concentrations typically depend on the specific research design Overall Pendimethalin remains a critical compound in herbicide research contributing to advancements in sustainable agricultural practices