Learn More
Cayman Chemical Pendimethalin, Purity: ≥98%, CAS: 40487-42-1, 50 mg
SDP
Catalog No. 502949254
Encompass_Preferred
A dinitroaniline herbicide; controls johnsongrass and increases seed cotton yield from 0.56 to 1.12 kg/ha; induces the production of ROS and genotoxicity in isolated human peripheral lymphocytes from 25-500 µM; increases renal and hepatic protein carbonylation, lipid peroxidation, necrosis, leukocyte infiltration, and formation of pyknotic nuclei in rats at 62.5, 125, and 250 mg/kg
Catalog No. 50-294-9254 Supplier Cayman Chemical Supplier No. 3455750 MG
A dinitroaniline herbicide; controls johnsongrass and increases seed cotton yield from 0.56 to 1.12 kg/ha; induces the production of ROS and genotoxicity in isolated human peripheral lymphocytes from 25-500 µM; increases renal and hepatic protein carbonylation, lipid peroxidation, necrosis, leukocyte infiltration, and formation of pyknotic nuclei in rats at 62.5, 125, and 250 mg/kg