Learn More
Medchemexpress LLC Pendimethalin-d5 | 1219803-39-0 | 99.9% | 1 MG
SDP
Catalog No. 5000409413
Encompass_Preferred
Pendimethalin-d5 is the deuterium labeled Pendimethalin. Pendimethalin is an herbicide that controls annual grasses and certain broadleaf weeds. It inhibits cell division and cell elongation.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-09413 Supplier Medchemexpress LLC Supplier No. HYB0862S1MG
Pendimethalin-d5 is the deuterium labeled Pendimethalin. Pendimethalin is an herbicide that controls annual grasses and certain broadleaf weeds. It inhibits cell division and cell elongation.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.