Learn More
Medchemexpress LLC Pendimethalin (Standard) | 40487-42-1 | 99.0% | 250 MG
SDP
Catalog No. 5000405607
Encompass_Preferred
Pendimethalin (Standard) is an analytical standard and an orally active herbicide. It controls annual grasses and certain broadleaf weeds by inhibiting cell division and cell elongation. It is also known to induce apoptotic cell death through activating ER stress-mediated mitochondrial dysfunction in human umbilical vein endothelial cells.
  • Intended for research and analytical applications.
  • Controls annual grasses and certain broadleaf weeds.
  • Used as an analytical standard in HPLC, GC, and MS research.
  • Induces apoptotic cell death through ER stress-mediated mitochondrial dysfunction.
Catalog No. 50-004-05607 Supplier Medchemexpress LLC Supplier No. 250MGHYB0862R
Add to Cart
Add to Cart
Pendimethalin (Standard) is an analytical standard and an orally active herbicide. It controls annual grasses and certain broadleaf weeds by inhibiting cell division and cell elongation. It is also known to induce apoptotic cell death through activating ER stress-mediated mitochondrial dysfunction in human umbilical vein endothelial cells.
  • Intended for research and analytical applications.
  • Controls annual grasses and certain broadleaf weeds.
  • Used as an analytical standard in HPLC, GC, and MS research.
  • Induces apoptotic cell death through ER stress-mediated mitochondrial dysfunction.