Learn More
Pendotech Pendotech Premium Flow Meter Monitor
Catalog No. NC1831881
Encompass
PendoTECH Premium Flow Meter Monitor with LCD and white backlit display; 4-20mA output, serial port, 12-24VDC power input range
Catalog No. NC1831881 Supplier Pendotech Supplier No. FMTS
PendoTECH Premium Flow Meter Monitor with LCD and white backlit display; 4-20mA output, serial port, 12-24VDC power input range