Learn More
BURKLE INC Pendulum beaker V2A, capacity 33.81 oz. (1000 ml), TeleScoop
Catalog No. 5000025878
Encompass_Preferred
This beaker automatically swings to a horizontal position. Nothing can be spilt. Suitable therefore for deep and narrow shafts (Ø 130 mm (5.12 in.)).
Catalog No. 50-000-25878 Supplier BURKLE INC Supplier No. 53543000
This beaker automatically swings to a horizontal position. Nothing can be spilt. Suitable therefore for deep and narrow shafts (Ø 130 mm (5.12 in.)).