Learn More
BURKLE INC Pendulum beaker PP, capacity 33.81 oz. (1000 ml), TeleScoop
Catalog No. 5000026051
Encompass_Preferred
This beaker automatically swings to a horizontal position. Nothing can be spilt. Suitable therefore for deep and narrow shafts (Ø 150 mm (5.91 in.)).
Catalog No. 50-000-26051 Supplier BURKLE INC Supplier No. 53545100
This beaker automatically swings to a horizontal position. Nothing can be spilt. Suitable therefore for deep and narrow shafts (Ø 150 mm (5.91 in.)).