Learn More
BURKLE INC Pendulum beaker PP, capacity 67.63 oz. (2000 ml), TeleScoop
Catalog No. 5000025697
Encompass_Preferred
This beaker automatically swings to a horizontal position. Nothing can be spilt. Suitable therefore for deep and narrow shafts (Ø 150 mm (5.91 in.)).
Catalog No. 50-000-25697 Supplier BURKLE INC Supplier No. 53545200
Add to Cart
Add to Cart
This beaker automatically swings to a horizontal position. Nothing can be spilt. Suitable therefore for deep and narrow shafts (Ø 150 mm (5.91 in.)).