Learn More
MSC Chute Lube, CRC Penetrant & Lubricant: 16 oz Spray Bottle
Supplier:  MSC 79280459
Encompass
Clear & White, 400 ° F Operating Temp, Flammable Non-Drying Film; Silicone; Voc Compliant
Catalog No. NC2748333