Learn More
Cole-Parmer PROBEK HI-TEMP -50/1000F
Supplier:  Cole-Parmer 9041507
Encompass
Special-application thermocouple probes work with any meter accepting standard thermocouple plug connectors. Choose the Type (J/K) compatible with your meter.
Catalog No. 50-993-112