Learn More
Perkin Elmer US LLC Penetration Probe Tip - Flat, 1 mm
Catalog No. 502111998
Encompass_Preferred
The Quartz Parallel Plate Systems are the same as the Stainless Steel Parallel Plate Measuring Systems, just made of quartz. These measuring systems ship in a attractive wooden case and include 1 and 6 mm parallel plate probes, mounting hardware and instructions. These systems are compatible with the DMA 7/7e and TMA analyzers.
Catalog No. 50-211-1998 Supplier Perkin Elmer US LLC Supplier No. N5190376
The Quartz Parallel Plate Systems are the same as the Stainless Steel Parallel Plate Measuring Systems, just made of quartz. These measuring systems ship in a attractive wooden case and include 1 and 6 mm parallel plate probes, mounting hardware and instructions. These systems are compatible with the DMA 7/7e and TMA analyzers.