Learn More
Cayman Chemical Penfluridol, Purity: ≥98%, CAS: 26864-56-2, 100 mg
SDP
Catalog No. 502936317
Encompass_Preferred
A typical antipsychotic; binds to dopamine receptors (IC50 = 5.6 nM); binds to dopamine D4 and 5-HT receptors (Kis = 31 and 63.1 nM, respectively); inhibits stereotypic behavior induced by apomorphine in cynomolgus monkeys at 1.25 and 2.5 mg/kg; inhibits proliferation of TNBCs (IC50s = 6-8 µM); inhibits migration and invasion of TNBCs and decreases the expression of integrin α6 and β4; reduces tumor growth and metastasis in a 4T1 orthotopic mouse model at 10 mg/kg per day
Catalog No. 50-293-6317 Supplier Cayman Chemical Supplier No. 26071100 MG
A typical antipsychotic; binds to dopamine receptors (IC50 = 5.6 nM); binds to dopamine D4 and 5-HT receptors (Kis = 31 and 63.1 nM, respectively); inhibits stereotypic behavior induced by apomorphine in cynomolgus monkeys at 1.25 and 2.5 mg/kg; inhibits proliferation of TNBCs (IC50s = 6-8 µM); inhibits migration and invasion of TNBCs and decreases the expression of integrin α6 and β4; reduces tumor growth and metastasis in a 4T1 orthotopic mouse model at 10 mg/kg per day