Learn More
Apexbio Technology LLC Penfluridol 26864-56-2 50mg
SDP
Catalog No. 502240820
Encompass_Preferred
Penfluridol (CAS 26864-56-2) is a first-generation antipsychotic belonging to the diphenylbutylpiperidine class It acts primarily as a dopamine D2 receptor antagonist disrupting dopaminergic neurotransmission in the central nervous system Penfluridol has been widely used in research to investigate dopaminergic signaling pathways and the pharmacology of antipsychotic agents Its molecular profile supports studies of receptor binding antipsychotic efficacy and the neurobiological basis of schizophrenia and related disorders
This item is not returnable. View return policy
Catalog No. 50-224-0820 Supplier Apexbio Technology LLC Supplier No. B181050
Penfluridol (CAS 26864-56-2) is a first-generation antipsychotic belonging to the diphenylbutylpiperidine class It acts primarily as a dopamine D2 receptor antagonist disrupting dopaminergic neurotransmission in the central nervous system Penfluridol has been widely used in research to investigate dopaminergic signaling pathways and the pharmacology of antipsychotic agents Its molecular profile supports studies of receptor binding antipsychotic efficacy and the neurobiological basis of schizophrenia and related disorders