Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penguins on Ice Math Activity Set
SCI_ED

Catalog No. s02210
Change view
Click to view available options
Grade:
K
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02210 K
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
CHOKING HAZARD–Small parts.
Catalog No. S02210 Supplier Learning Resources, Inc Supplier No. LER3311
Only null left
Add to Cart
Add to Cart
CHOKING HAZARD–Small parts.

Includes

  • 10 plastic ice bars, 100 plastic penguins in 10 colors, and Activity Guide
  • CHOKING HAZARD (1). Not for under 3 yr.

Cool penguin set goes the extra mile—so versatile!

  • Ice bars hold 10 penguins each and connect two ways—horizontally (number line) or vertically (hundred grid)
  • Reinforces math concepts—numbers, counting, patterning, sorting, operations, hundreds and base ten
  • 12L x 1 in.H (30.48 x 2.54cm)

Order Info

Grades K+

Specifications

Color Assorted
Grade Level Elementary
Grade K
For Use With (Application) Numbers, counting, patterning, sorting, operations, hundreds and base ten
Includes 10 plastic ice bars, 100 plastic penguins in 10 colors, and activity guide
Description Penguins on Ice Math Activity Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.