Learn More
Cayman Chemical Penicillamine-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2925655-01-0, 1 mg
SDP
Catalog No. 502949384
Encompass_Preferred
An internal standard for the quantification of penicillamine by GC- or LC-MS
Catalog No. 50-294-9384 Supplier Cayman Chemical Supplier No. 337971 MG
An internal standard for the quantification of penicillamine by GC- or LC-MS