Learn More
Cayman Chemical PenIcIlIc AcId 10mg
SDP
Catalog No. 502953072
Encompass_Preferred
A mycotoxin produced from Penicillium species that inhibits bacterial quorum sensing communication in P. aeruginosa; inhibits Fas-mediated apoptosis (100-200 μM) by targeting caspase-8 activity in Burkitt’s lymphoma Raji cells
Catalog No. 50-295-3072 Supplier Cayman Chemical Supplier No. 1144010 MG
A mycotoxin produced from Penicillium species that inhibits bacterial quorum sensing communication in P. aeruginosa; inhibits Fas-mediated apoptosis (100-200 μM) by targeting caspase-8 activity in Burkitt’s lymphoma Raji cells