Learn More
Cayman Chemical PenIcIlIn GpotasIum salt 25g
SDP
Catalog No. 502987505
Encompass_Preferred
A β-lactam antibiotic; effective mainly for Gram-positive bacteria; MIC90s = 0.03, 1, and 4 UG/ml for penicillin-susceptible, -intermediate, and -resistant strains of S. pneumoniae; MIC90s = 16-128 UG/ml for various Bacteroides isolates in a broth dilution assay
Catalog No. 50-298-7505 Supplier Cayman Chemical Supplier No. 2161525 G
A β-lactam antibiotic; effective mainly for Gram-positive bacteria; MIC90s = 0.03, 1, and 4 UG/ml for penicillin-susceptible, -intermediate, and -resistant strains of S. pneumoniae; MIC90s = 16-128 UG/ml for various Bacteroides isolates in a broth dilution assay