Learn More
Supelco Inc PENICILIN STREPTOMINE 100 ML
Catalog No. NC9778496
Encompass
Penicillin Aˆ' Streptomycin Aˆ' Neomycin Solution StAbilized, formulAted to contAin ~5,000 units penicillin, 5 mg streptomycin And 10 mg neomycin/mL, sterile-filtered, BioReAgent, suitAble for cell culture
Catalog No. NC9778496 Supplier Supelco Inc Supplier No. P4083100ML
Penicillin Aˆ' Streptomycin Aˆ' Neomycin Solution StAbilized, formulAted to contAin ~5,000 units penicillin, 5 mg streptomycin And 10 mg neomycin/mL, sterile-filtered, BioReAgent, suitAble for cell culture