Learn More
Cayman Chemical Bilaid A, Purity: ≥95%, CAS: 2393865-97-7, 5 mg
SDP
Catalog No. 502991555
Encompass_Preferred
A tetrapeptide μ-opioid receptor agonist (Ki = 3.1 μM using HEK293 cell membranes expressing the human receptor); inhibits forskolin-induced cAMP accumulation in HEK293cells expressing the human μ-opioid receptor by 21% at 10 μM
Catalog No. 50-299-1555 Supplier Cayman Chemical Supplier No. 312425 MG
A tetrapeptide μ-opioid receptor agonist (Ki = 3.1 μM using HEK293 cell membranes expressing the human receptor); inhibits forskolin-induced cAMP accumulation in HEK293cells expressing the human μ-opioid receptor by 21% at 10 μM