Learn More
Cayman Chemical Cyclopenin, Purity: ≥98%, CAS: 20007-87-8, 5 mg
SDP
Catalog No. 502995083
Encompass_Preferred
An AChE inhibitor (IC50 = 2.04 μM for human recombinant AChE); exhibits greater than 2,000-fold selectivity over equine butyrylcholinesterase (IC50 = >4,080 μM)
Catalog No. 50-299-5083 Supplier Cayman Chemical Supplier No. 231455 MG
An AChE inhibitor (IC50 = 2.04 μM for human recombinant AChE); exhibits greater than 2,000-fold selectivity over equine butyrylcholinesterase (IC50 = >4,080 μM)