Learn More
Cayman Chemical NG 012, Purity: ≥95%, CAS: 141731-76-2, 5 mg
SDP
Catalog No. 502941560
Encompass_Preferred
A fungal metabolite; inhibits the growth of the plant pathogenic fungi B. cinerea, P. teres, P. lingam, S. sclerotiorum, and M. fructigena in vitro; increases the efficacy of miconazole against C. albicans, reducing the IC50 value from 19.2 to 2.5 μM when used at 50 μM; potentiates NGF-induced neurite outgrowth in PC12 cells in a concentration-dependent manner
Catalog No. 50-294-1560 Supplier Cayman Chemical Supplier No. 275095 MG
A fungal metabolite; inhibits the growth of the plant pathogenic fungi B. cinerea, P. teres, P. lingam, S. sclerotiorum, and M. fructigena in vitro; increases the efficacy of miconazole against C. albicans, reducing the IC50 value from 19.2 to 2.5 μM when used at 50 μM; potentiates NGF-induced neurite outgrowth in PC12 cells in a concentration-dependent manner