Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Penicillic acid | 90-65-3 | 10 MG
SDP

Catalog No. 5000419309
Encompass_Preferred

Penicillic acid is a polyketide mycotoxin produced by several species of Aspergillus and Penicillium. It exhibits cytotoxicity in rat alveolar macrophages in vitro and inhibits Fas ligand-induced apoptosis by blocking the self-processing of caspase-8.

  • Polyketide mycotoxin
  • Exhibits cytotoxicity in rat alveolar macrophages
  • Inhibits Fas ligand-induced apoptosis
  • Blocks self-processing of caspase-8
  • Purity: 99.63%
  • Appearance: Solid
  • Color: White to off-white
  • Soluble in DMSO (100 mg/mL)
  • Soluble in H2O (25 mg/mL)
  • Inhibits DNA fragmentation induced by Jo2
  • Reverses reduced cell viability exposed to Jo2
  • Prevents caspase-8 processing in Raji cells

Catalog No. 50-004-19309 Supplier Medchemexpress LLC Supplier No. HYN677710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Penicillic acid is a polyketide mycotoxin produced by several species of Aspergillus and Penicillium. It exhibits cytotoxicity in rat alveolar macrophages in vitro and inhibits Fas ligand-induced apoptosis by blocking the self-processing of caspase-8.

  • Polyketide mycotoxin
  • Exhibits cytotoxicity in rat alveolar macrophages
  • Inhibits Fas ligand-induced apoptosis
  • Blocks self-processing of caspase-8
  • Purity: 99.63%
  • Appearance: Solid
  • Color: White to off-white
  • Soluble in DMSO (100 mg/mL)
  • Soluble in H2O (25 mg/mL)
  • Inhibits DNA fragmentation induced by Jo2
  • Reverses reduced cell viability exposed to Jo2
  • Prevents caspase-8 processing in Raji cells

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.