Learn More
AdipoGen Penicillide (1 mg)
SDP
Supplier:  AdipoGen AGCN20122M001
Encompass_Preferred
Chemical. CAS: 55303-92-9. Formula: C21H24O6. MW: 372.4. Isolated from Penicillium sp. Plant growth inhibitor. Acyl-CoA-cholesterol acyltransferase (ACAT) inhibitor. Anticancer compound. Cytotoxic. Non-peptidic oxytocin receptor antagonist. Calpain inhibitor.
Catalog No. 50-205-3491