Learn More
Medchemexpress LLC Penicillin amidase, E. coli | 9014-06-6 | 100 MG
SDP
Catalog No. 5000400296
Encompass_Preferred
Penicillin amidase, E. coli (Immobilized) (PGA) is an amidohydrolase commonly used in industrial biocatalysis. It serves as a starting material in the synthesis of semi-synthetic penicillins and promotes the production of semi-synthetic β-lactam antibiotics.
  • Participates in peptide synthesis
  • Catalyzes the formation of chiral compounds
  • Activity regulated by temperature and phenylacetic acid in E. coli
  • Involved in assimilation of aromatic compounds as a carbon source in free E. coli
  • Hypothesized to act as a scavenging enzyme for phenylacetyl-containing compounds in microbial metabolism
  • Associated with bacterial quorum sensing
Catalog No. 50-004-00296 Supplier Medchemexpress LLC Supplier No. HYP2834100MG
Penicillin amidase, E. coli (Immobilized) (PGA) is an amidohydrolase commonly used in industrial biocatalysis. It serves as a starting material in the synthesis of semi-synthetic penicillins and promotes the production of semi-synthetic β-lactam antibiotics.
  • Participates in peptide synthesis
  • Catalyzes the formation of chiral compounds
  • Activity regulated by temperature and phenylacetic acid in E. coli
  • Involved in assimilation of aromatic compounds as a carbon source in free E. coli
  • Hypothesized to act as a scavenging enzyme for phenylacetyl-containing compounds in microbial metabolism
  • Associated with bacterial quorum sensing