Learn More
Medchemexpress LLC Penicillin amidase, E. coli | 9014-06-6 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYP2834
Encompass_Preferred
Penicillin amidase, E. coli (Immobilized) (PGA) is an amidohydrolase commonly used in industrial biocatalysis. It serves as a starting material in the synthesis of semi-synthetic penicillins and promotes the production of semi-synthetic β-lactam antibiotics. It also participates in peptide synthesis and catalyzes the formation of chiral compounds.
  • Used in industrial biocatalysis.
  • Serves as a starting material in semi-synthetic penicillin synthesis.
  • Promotes production of semi-synthetic β-lactam antibiotics.
  • Participates in peptide synthesis.
  • Catalyzes the formation of chiral compounds.
  • Regulated by temperature and phenylacetic acid in E. coli.
  • Assimilates aromatic compounds as a carbon source in free E. coli.
  • Hypothesized to act as a scavenging enzyme for phenylacetyl-containing compounds.
  • Associated with bacterial quorum sensing.
Catalog No. 50-003-97203