Learn More
MOLECULAR DEPOT LLC Penicillin G BSA - 1 mg
Catalog No. 5000013517
Encompass_Preferred
Penicillin G BSA conjugate is formed by linking Penicillin G to Bovine Serum Albumin (BSA) BSA serves as a stabilizer and carrier protein in laboratory experiments This conjugate aids in understanding immune recognition of penicillin For in vitro research use only
Catalog No. 50-000-13517 Supplier MOLECULAR DEPOT LLC Supplier No. B2020759
Penicillin G BSA conjugate is formed by linking Penicillin G to Bovine Serum Albumin (BSA) BSA serves as a stabilizer and carrier protein in laboratory experiments This conjugate aids in understanding immune recognition of penicillin For in vitro research use only