Learn More
Medchemexpress LLC Penicillin G-d5 potassium (Benzylpenicillin-d5 potassium) | 113-98-4 | 98.7% | 377.51 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY17591S5MG
Encompass_Preferred
Penicillin G-d5 (potassium) is the deuterium labeled Penicillin G potassium. Penicillin G potassium is a fast-acting antibiotic used to treat bacterial infections that affect the blood, heart, lungs, joints, and genital areas. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-87369