Learn More
Med Vet International Penicillin G Procaine, Injectable Susp, 100mL, 300, 000 Units, Vet Label
Catalog No. 502828838
Encompass_Preferred
VetriPen G (penicillin G procaine 300, 000 units – Reduced Withdrawal Time) is an effective bactericide in the treatment of infections caused primarily by penicillin-sensitive organisms, such as Streptococcus equi and Erysipelothrix rhusiopathiae, as well as the gram negative organism Pasteurella multocida. It is recommended for treatment of bacterial pneumonia (shipping fever) caused by Pasteurella multocida in cattle and sheep, erysipelas caused by Erysipelothrix insidiosa in swine, and strangles caused by Streptococcus equi in horses. Milk withhold: 48 hours. Meat withdrawal: Cattle 4 days, Sheep 8 days, Swine 6 days, Non-Ruminating Calves - 7 days. Administer IM. Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet can only ship cold items Monday-Wednesday. *Next Day Air Available With Additional Charges* NOT FOR SALE IN CALIFORNIA
Catalog No. 50-282-8838 Supplier Med Vet International Supplier No. RXPENG100
VetriPen G (penicillin G procaine 300, 000 units – Reduced Withdrawal Time) is an effective bactericide in the treatment of infections caused primarily by penicillin-sensitive organisms, such as Streptococcus equi and Erysipelothrix rhusiopathiae, as well as the gram negative organism Pasteurella multocida. It is recommended for treatment of bacterial pneumonia (shipping fever) caused by Pasteurella multocida in cattle and sheep, erysipelas caused by Erysipelothrix insidiosa in swine, and strangles caused by Streptococcus equi in horses. Milk withhold: 48 hours. Meat withdrawal: Cattle 4 days, Sheep 8 days, Swine 6 days, Non-Ruminating Calves - 7 days. Administer IM. Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet can only ship cold items Monday-Wednesday. *Next Day Air Available With Additional Charges* NOT FOR SALE IN CALIFORNIA