Learn More
Research Products International Corp Penicillin G Potasm Salt 500 G
SDP
Catalog No. 502230541
Encompass_Preferred
Penicillin G potassium salt is a member of the penicillin class of antibiotics and is effective against a variety of gram-positive bacteria. As a narrow-spectrum antibiotic belonging to the beta-lactam class, it inhibits the synthesis of bacterial cell walls. It interferes with the formation of peptidoglycan, a crucial component of the bacterial cell wall. This weakens the cell wall and leads to the eventual rupture of the bacterial cell.
Penicillin G is effective against a wide range of Gram-positive bacteria, including Streptococcus, Staphylococcus, and some other species. However, it is not effective against Gram-negative bacteria due to their different cell wall structure.
Catalog No. 50-223-0541 Supplier Research Products International Corp Supplier No. P93500500.0
Penicillin G potassium salt is a member of the penicillin class of antibiotics and is effective against a variety of gram-positive bacteria. As a narrow-spectrum antibiotic belonging to the beta-lactam class, it inhibits the synthesis of bacterial cell walls. It interferes with the formation of peptidoglycan, a crucial component of the bacterial cell wall. This weakens the cell wall and leads to the eventual rupture of the bacterial cell.
Penicillin G is effective against a wide range of Gram-positive bacteria, including Streptococcus, Staphylococcus, and some other species. However, it is not effective against Gram-negative bacteria due to their different cell wall structure.