Learn More
Medchemexpress LLC Penicillin G (potassium) | 113-98-4 | 99.6% | 372.48 | 10 G
SDP
Catalog No. 5000387509
Encompass_Preferred
Penicillin G potassium is a fast-acting penicillin family antibiotic used for the research of bacterial infections that affect the blood, heart, lungs, joints, and genital areas.
  • Fast-acting penicillin family antibiotic.
  • Can be used for research of bacterial infections.
  • Effective against _E. coli_ in combination with promethazine in vitro.
Catalog No. 50-003-87509 Supplier Medchemexpress LLC Supplier No. HY1759110G
Penicillin G potassium is a fast-acting penicillin family antibiotic used for the research of bacterial infections that affect the blood, heart, lungs, joints, and genital areas.
  • Fast-acting penicillin family antibiotic.
  • Can be used for research of bacterial infections.
  • Effective against _E. coli_ in combination with promethazine in vitro.