Learn More
Medchemexpress LLC Penicillin G (potassium) | 113-98-4 | 99.6% | 372.48 | 100 G
SDP
Catalog No. 5000387094
Encompass_Preferred
Penicillin G potassium is a fast-acting penicillin family antibiotic. It is used for the research of bacterial infections that affect the blood, heart, lungs, joints, and genital areas. It is suitable for laboratory centrifugation and storage.
  • Suitable for research of bacterial infections
  • Fast-acting penicillin family antibiotic
  • Maintained at 4°C, sealed storage, away from moisture
  • Compound is unstable in solutions, freshly prepared is recommended
Catalog No. 50-003-87094 Supplier Medchemexpress LLC Supplier No. HY17591100G
Penicillin G potassium is a fast-acting penicillin family antibiotic. It is used for the research of bacterial infections that affect the blood, heart, lungs, joints, and genital areas. It is suitable for laboratory centrifugation and storage.
  • Suitable for research of bacterial infections
  • Fast-acting penicillin family antibiotic
  • Maintained at 4°C, sealed storage, away from moisture
  • Compound is unstable in solutions, freshly prepared is recommended