Learn More
Medchemexpress LLC Penicillin G potassium | 113-98-4 | 99.7% | 372.48 | 25 G
SDP
Catalog No. 5000385549
Encompass_Preferred
Penicillin G potassium is a fast-acting penicillin family antibiotic. It can be used for the research of bacterial infections that affect the blood, heart, lungs, joints, and genital areas.
  • Solid appearance
  • White to off-white color
  • Classified as an antibiotic for disease research, antibacterial, animal husbandry, plant cell culture, and mammalian cell culture
  • Initially sourced from microorganisms, Penicillium
  • Ships at room temperature in continental US
  • Store at 4°C, sealed, away from moisture
Catalog No. 50-003-85549 Supplier Medchemexpress LLC Supplier No. 25GHY17591
Penicillin G potassium is a fast-acting penicillin family antibiotic. It can be used for the research of bacterial infections that affect the blood, heart, lungs, joints, and genital areas.
  • Solid appearance
  • White to off-white color
  • Classified as an antibiotic for disease research, antibacterial, animal husbandry, plant cell culture, and mammalian cell culture
  • Initially sourced from microorganisms, Penicillium
  • Ships at room temperature in continental US
  • Store at 4°C, sealed, away from moisture