Learn More
Alkali Scientific Penicillin G Potassium Salt, 500 G
SDP
Catalog No. 5000660188
Encompass_Preferred
Penicillin G Potassium Salt is a potent antibiotic widely used in microbiological research and clinical applications to combat infections caused by gram-positive bacteria. The 500 g powdered formulation is ideal for large-scale laboratory applications, where Penicillin G is needed in high concentrations. It is used to prepare solutions for antimicrobial susceptibility testing, bacterial growth inhibition studies, and resistance research. The compound works by interfering with bacterial cell wall synthesis, leading to bacterial cell death. Penicillin G Potassium is a valuable tool for studying the effectiveness of antibiotics in treating resistant bacterial strains and for testing new antimicrobial compounds in a laboratory setting.
Catalog No. 50-006-60188 Supplier Alkali Scientific Supplier No. ENI003CP
Penicillin G Potassium Salt is a potent antibiotic widely used in microbiological research and clinical applications to combat infections caused by gram-positive bacteria. The 500 g powdered formulation is ideal for large-scale laboratory applications, where Penicillin G is needed in high concentrations. It is used to prepare solutions for antimicrobial susceptibility testing, bacterial growth inhibition studies, and resistance research. The compound works by interfering with bacterial cell wall synthesis, leading to bacterial cell death. Penicillin G Potassium is a valuable tool for studying the effectiveness of antibiotics in treating resistant bacterial strains and for testing new antimicrobial compounds in a laboratory setting.