Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penicillin G potassium salt, 99.9%, MP Biomedicals™

Catalog No. ICN10054380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mU
10 mU
100 mU
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10054380 100 mU
ICN10054301 1 mU
ICN10054310 10 mU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10054380 Supplier MP Biomedicals, Inc Supplier No. 0210054380
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 113-98-4
Molecular Formula C33H36ClKN4O4S2
Molecular Weight (g/mol) 691.34
MDL Number MFCD00036193
InChI Key HTQQVOYPSGNVPA-HBRCYENSSA-M
Synonym Benzylpenicillin potassium salt
PubChem CID 23664709
ChEBI CHEBI:7963
IUPAC Name potassium [3-(2-chloro-10H-phenothiazin-10-yl)propyl]dimethylamine (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [K+].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(Cl)C=C12.CC1(C)S[C@@H]2[C@H](NC(=O)CC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O

Specifications

Type Penicillin G Potassium Salt
Physical Form Powder
Color White
Quantity 100 mU
Assay Percent Range 0.999
Formula Weight 372.5
Grade Not Available
Chemical Name or Material Penicillin G potassium salt, Potency: 1500-1700 u/mg
EINECSNumber 204-038-0
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.