Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Hydroxyquinoline, 99%

Catalog No. BP914100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP914-100 100 mU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BP914-100

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 113-98-4 | C33H36ClKN4O4S2 | 691.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

  • Penicillin G is a narrow spectrum antibiotic that is effective against gram positive bacteria
  • Useful for preventing microbial contamination in tissue culture applications
  • Functions as an antiobiotic by inhibiting cell wall synthesis 
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Specifications

Type Penicillin G Potassium Salt
Chemical Name or Material Penicillin-G Potassium Salt
Physical Form Solid
CAS 113-98-4
pH 5.5 to 7.5
Melting Point 214°C
Loss on Drying < 1.5%
Packaging Poly Bottle
Potency Inclusive between 1440μg/mg
Color White
Quantity 100 mU
Assay Percent Range ≥85 %
Molecular Formula C33H36ClKN4O4S2
MDL Number MFCD00036193
Merck Index 15, 7203
Synonym Benzylpenicillin potassium salt
InChI Key HTQQVOYPSGNVPA-HBRCYENSSA-M
SMILES [K+].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(Cl)C=C12.CC1(C)S[C@@H]2[C@H](NC(=O)CC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
IUPAC Name potassium;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol) 691.34
PubChem CID 23664709
ChEBI CHEBI:7963
Formula Weight 372.47
Percent Purity ≥85%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.