Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penicillin G Potassium Salt , MP Biomedical

Catalog No. MP210054310 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210054310 10 mU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210054310 Supplier MP Biomedicals, Inc Supplier No. 0210054310
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Penicillin G Potassium Salt is a narrow spectrum antibiotic. It is the drug of choice for Streptococcus pneumoniae, groups A, B, C and G streptococci, nonenterococcal group D streptococci, viridans group streptococci, and non-penicillinase producing staphylococcus.

  • Penicillin G potassium salt is used to study murosomes of staphylococci and the penicillin-induced lysis of Streptococcus mutants
  • Active against gram-positive and gram-negative aerobic and anaerobic bacteria
  • Inhibits cell wall synthesis by binding to penicillin binding proteins which inhibit peptidoglycan chain cross-linking
  • Stable against hydrolysis by β-lactamases, such as penicillinases and cephalosporinases
TRUSTED_SUSTAINABILITY

Specifications

pH 5 to 7.5 (3% aq. soln.)
Melting Point 214°C to 217°C (literature)
Packaging Amber Glass Bottle
Color White
Quantity 10 mU
Percent Purity 99.8%
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.