Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penicillin G Potassium Salt, MP Biomedicals™

Catalog No. MP210054325 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210054325 25 mU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210054325 Supplier MP Biomedicals, Inc Supplier No. 0210054325
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Active against gram-positive and gram-negative aerobic and anaerobic bacteria. Penicillin G Potassium Salt, MP Biomedicals is a narrow spectrum antibiotic. It inhibits cell wall synthesis by binding to penicillin binding proteins (PBPs), which inhibits pe

Penicillin G is the drug of choice for Streptococcus pneumoniae, groups A, B, C and G streptococci, nonenterococcal group D streptococci, viridans group streptococci, and non-penicillinase producing staphylococcus.

Penicillin G potassium salt is used to study murosomes of staphylococci and the penicillin-induced lysis of Streptococcus mutans. It is stable against hydrolysis by β-lactamases, such as penicillinases and cephalosporinases.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113-98-4
Molecular Formula C33H36ClKN4O4S2
Molecular Weight (g/mol) 691.34
MDL Number MFCD00036193
InChI Key HTQQVOYPSGNVPA-HBRCYENSSA-M
Synonym Benzylpenicillin potassium salt
PubChem CID 23664709
ChEBI CHEBI:7963
IUPAC Name potassium [3-(2-chloro-10H-phenothiazin-10-yl)propyl]dimethylamine (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [K+].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(Cl)C=C12.CC1(C)S[C@@H]2[C@H](NC(=O)CC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O

Specifications

Type Penicillin G Potassium Salt
Physical Form Powder
pH 5 to 7.5 (3% aq. soln.)
Melting Point 214°C to 217°C (literature)
Quantity 25 mU
Solubility Information Soluble in water (100mg/mL) clear,colorless to light yellow.
Formula Weight 372.48
Grade Not Available
Recommended Storage Store at 4°C
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.