Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penicillin G Potassium Salt, USP Grade, G-Biosciences
SCI_ED

Catalog No. S139815
Change view
Click to view available options
Quantity:
500 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139815 500 g
S000035 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139815 Supplier G-Biosciences Supplier No. RC863
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Penicillin G is a naturally occurring beta lactam antibiotic produced by the Penicillium fungus. Tested against both sensitive and resistant cells.

Penicillin G Potassium works by inhibiting cell wall production in non-resistant cells by breaking the links between peptidogylocans and causing the production of cell wall damaging autolysins.

Chemical Identifiers

CAS 113-98-4
Molecular Weight (g/mol) 372.48 g/mol

Specifications

Physical Form Flakes or Powder
Quantity 500 g
Percent Purity >98%
Grade USP
Chemical Name or Material Penicillin G Potassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.