Learn More
Medchemexpress LLC Penicillin G (potassium) (Standard) | 113-98-4 | 99.9% | 372.48 | 25 MG
SDP
Catalog No. 5000384740
Encompass_Preferred
Penicillin G (potassium) (Standard) is the analytical standard of Penicillin G (potassium), intended for research and analytical applications. It is a fast-acting penicillin family antibiotic used for the research of bacterial infections that affect the blood, heart, lungs, joints, and genital areas. This analytical standard is commonly employed in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for research applications
  • Fast-acting penicillin family antibiotic
  • Suitable for bacterial infection research
  • Used in HPLC, GC, and MS experiments
Catalog No. 50-003-84740 Supplier Medchemexpress LLC Supplier No. 25MGHY17591R
Penicillin G (potassium) (Standard) is the analytical standard of Penicillin G (potassium), intended for research and analytical applications. It is a fast-acting penicillin family antibiotic used for the research of bacterial infections that affect the blood, heart, lungs, joints, and genital areas. This analytical standard is commonly employed in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for research applications
  • Fast-acting penicillin family antibiotic
  • Suitable for bacterial infection research
  • Used in HPLC, GC, and MS experiments