Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Penicillin G procaine hydrate | 6130-64-9 | 1 ML
SDP

Catalog No. 5000421099
Encompass_Preferred

Penicillin G Procaine hydrate (PGP hydrate) is a β-lactam antibiotic and a sustained-release formulation. In vivo, Penicillin G procaine (hydrate) hydrolyzes to release penicillin G and procaine, which delays the absorption of penicillin G.

  • DMSO solubility: 250 mg/mL (424.65 mM); requires ultrasonic; use newly opened DMSO due to hygroscopic nature affecting solubility
  • H2O solubility: 3.14 mg/mL (5.33 mM); requires ultrasonic
  • Powder storage: -20°C for 3 years or 4°C for 2 years
  • In solvent (stock solution) storage: -80°C for 2 years or -20°C for 1 year
  • Aliquot and store prepared solutions to prevent inactivation from repeated freeze-thaw cycles

Catalog No. 50-004-21099 Supplier Medchemexpress LLC Supplier No. HYN712010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Penicillin G Procaine hydrate (PGP hydrate) is a β-lactam antibiotic and a sustained-release formulation. In vivo, Penicillin G procaine (hydrate) hydrolyzes to release penicillin G and procaine, which delays the absorption of penicillin G.

  • DMSO solubility: 250 mg/mL (424.65 mM); requires ultrasonic; use newly opened DMSO due to hygroscopic nature affecting solubility
  • H2O solubility: 3.14 mg/mL (5.33 mM); requires ultrasonic
  • Powder storage: -20°C for 3 years or 4°C for 2 years
  • In solvent (stock solution) storage: -80°C for 2 years or -20°C for 1 year
  • Aliquot and store prepared solutions to prevent inactivation from repeated freeze-thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.