Learn More
TARGETMOL CHEMICALS INC Penicillin G sodium salt 500MG
SDP
Catalog No. 502880752
Encompass_Preferred
Also available in 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Penicillin G sodium(Benzylpenicillin sodium) salt is a penicillin derivative commonly used in the form of its sodium or potassium salts in the treatment of a variety of infections. It is effective against most gram-positive bacteria and against gram-negative cocci. It has also been used as an experimental convulsant because of its actions on GAMMA-AMINOBUTYRIC ACID mediated synaptic transmission. Purity 98.11%
Catalog No. 50-288-0752 Supplier TARGETMOL CHEMICALS INC Supplier No. T0958500MG
Also available in 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Penicillin G sodium(Benzylpenicillin sodium) salt is a penicillin derivative commonly used in the form of its sodium or potassium salts in the treatment of a variety of infections. It is effective against most gram-positive bacteria and against gram-negative cocci. It has also been used as an experimental convulsant because of its actions on GAMMA-AMINOBUTYRIC ACID mediated synaptic transmission. Purity 98.11%